Gene Transcripts

Gene Symbol
IL6R
Entrez Gene ID
3570
Gene Name
interleukin 6 receptor
Gene Aliases
CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
Chromosome Location
Chr.1: 154405193 - 154469450 on Build GRCh38
Uni Gene
Hs.135087
Interrogated Sequence Translated Protein
RefSeq NM_000565.3 NP_000556.1
NM_001206866.1 NP_001193795.1
NM_181359.2 NP_852004.1
XM_005245139.1 XP_005245196.1
XM_005245140.2 XP_005245197.1
XM_006711298.1 XP_006711361.1
XM_006711299.3 XP_006711362.1
XM_017001199.1 XP_016856688.1
XM_017001200.1 XP_016856689.1
XM_017001201.1 XP_016856690.1
GenBank mRNA AA035377.1 -
AK223582.1 -
AK293013.1 -
AK312730.1 -
BC035327.1 -
BC089410.1 -
BC132684.1 -
BC132686.1 -
BM911928.1 -
S72848.1 -
X12830.1 -
X58298.1 -

View Full Product Details