Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000142.4 | NP_000133.1 |
NM_001163213.1 | NP_001156685.1 | |
NM_022965.3 | NP_075254.1 | |
XM_006713868.1 | XP_006713931.1 | |
XM_006713869.1 | XP_006713932.1 | |
XM_006713870.1 | XP_006713933.1 | |
XM_006713871.1 | XP_006713934.1 | |
XM_006713872.1 | XP_006713935.1 | |
XM_006713873.1 | XP_006713936.1 | |
XM_011513420.1 | XP_011511722.1 | |
XM_011513422.1 | XP_011511724.1 | |
GenBank mRNA | AB209441.1 | - |
AF238374.1 | - | |
AF245114.1 | - | |
AF369211.1 | - | |
AF369212.1 | - | |
AF369213.1 | - | |
AK308936.1 | - | |
AW204106.1 | - | |
BC121175.2 | - | |
BC128610.1 | - | |
BC153824.1 | - | |
DR003490.1 | - | |
ENST00000440486.0 | - | |
HM244683.1 | - | |
KJ534834.1 | - | |
KJ534974.1 | - | |
KJ535004.1 | - | |
M58051.1 | - | |
M59374.1 | - | |
M64347.1 | - | |
X84939.1 | - |