Gene Transcripts

Gene Symbol
Flt4
Entrez Gene ID
14257
Gene Name
FMS-like tyrosine kinase 4
Gene Aliases
AI323512, Chy, Flt-4, VEGFR-3, VEGFR3
Chromosome Location
Chr.11: 49609679 - 49652739 on Build GRCm38
Uni Gene
Mm.3291
Interrogated Sequence Translated Protein
RefSeq NM_008029.3 NP_032055.1
GenBank mRNA AK090148.1 -
BC138346.1 -
BC138348.1 -
BU757635.1 -
BY311090.1 -
L07296.1 -

View Full Product Details