Gene Transcripts

Gene Symbol
Ccl25
Entrez Gene ID
20300
Gene Name
chemokine (C-C motif) ligand 25
Gene Aliases
A130072A22Rik, AI852536, CKb15, Scya25, TECK
Chromosome Location
Chr.8: 4325159 - 4360020 on Build GRCm38
Uni Gene
Mm.7275
Interrogated Sequence Translated Protein
RefSeq NM_009138.3 NP_033164.1
NR_033527.1
GenBank mRNA AJ249480.1 -
AK038019.1 -
AK043043.1 -
AK086889.1 -
AK131875.1 -
AK132700.1 -
AK134138.1 -
AK154211.1 -
BC117033.1 -
BC117037.1 -
DQ158256.1 -
U86357.1 -

View Full Product Details