Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_009138.3 | NP_033164.1 |
NR_033527.1 | ||
GenBank mRNA | AJ249480.1 | - |
AK038019.1 | - | |
AK043043.1 | - | |
AK086889.1 | - | |
AK131875.1 | - | |
AK132700.1 | - | |
AK134138.1 | - | |
AK154211.1 | - | |
BC117033.1 | - | |
BC117037.1 | - | |
DQ158256.1 | - | |
U86357.1 | - |