Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001277932.1 | NP_001264861.1 |
NM_015733.5 | NP_056548.2 | |
GenBank mRNA | AB019600.1 | - |
AB019601.1 | - | |
AK085095.1 | - | |
AK085152.1 | - | |
AK085561.1 | - | |
AK154495.1 | - | |
AK156500.1 | - | |
AK162863.1 | - | |
AK203861.1 | - | |
BC056372.1 | - | |
BC056447.1 | - | |
BY549141.1 | - | |
CT010400.1 | - |