Gene Transcripts

Gene Symbol
Il6r
Entrez Gene ID
24499
Gene Name
interleukin 6 receptor
Gene Aliases
IL6R1, Il6ra
Chromosome Location
Chr.2: 189196338 - 189255986 on Build Rnor_6.0
Uni Gene
Rn.1716
Interrogated Sequence Translated Protein
RefSeq NM_017020.3 NP_058716.2
XM_006232587.2 XP_006232649.1
XM_006232588.2 XP_006232650.1
XM_008761127.1 XP_008759349.1
GenBank mRNA CB778659.1 -
M58587.1 -

View Full Product Details