Gene Transcripts
- Gene Symbol
- Copg2os2
- Entrez Gene ID
- 100044236
- Gene Name
- coatomer protein complex, subunit gamma 2, opposite strand 2
- Gene Aliases
- AW049845, Copg2as2, Lb9, Mit1
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.22891
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002845.1 |
|
|
1234 |
|
GenBank mRNA |
AK045748.1 |
- |
|
460 |
|
U20264.1 |
- |
|
1234 |
|
View Full Product Details