Gene Transcripts
- Gene Symbol
- 2210417A02Rik
- Entrez Gene ID
- 70138
- Gene Name
- RIKEN cDNA 2210417A02 gene
- Gene Aliases
- AV081004
- Chromosome Location
- Chr. 5: 148741840 - 148743139 on Build GRCm38
- UniGene ID
- Mm.417834
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028285.1 |
|
2 |
269 |
|
GenBank mRNA |
AK008960.1 |
- |
2 |
268 |
|
View Full Product Details