Gene Transcripts
- Gene Symbol
- C330024D21Rik
- Entrez Gene ID
- 320479
- Gene Name
- RIKEN cDNA C330024D21 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 67463898 - 67477899 on Build GRCm38
- UniGene ID
- Mm.387187
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015582.2 |
|
5 |
1517 |
|
GenBank mRNA |
AK049327.1 |
- |
5 |
1518 |
|
View Full Product Details