Gene Transcripts
- Gene Symbol
- ZNF503-AS2
- Entrez Gene ID
- 100131213
- Gene Name
- ZNF503 antisense RNA 2
- Gene Aliases
- C10orf41, NCRNA00245
- Chromosome Location
- Chr. 10: 75401528 - 75408982 on Build GRCh38
- UniGene ID
- Hs.534598
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA135297.1 |
- |
2 |
504 |
|
Chr. 10 - 75402045 - 75407589 [+] |
GenBankmRNA |
BX344557.2 |
- |
2 |
650 |
|
Chr. 10 - 75401898 - 75407941 [+] |
GenBankmRNA |
BX400778.2 |
- |
2 |
650 |
|
Chr. 10 - 75401898 - 75407779 [+] |
GenBankmRNA |
BF343288.1 |
- |
|
415 |
|
Chr. - - [-] |
View Full Product Details