Gene Transcripts

Gene Symbol
KLRA1P
Entrez Gene ID
10748
Gene Name
killer cell lectin like receptor A1, pseudogene
Gene Aliases
KLRA1, KLRAP1, LY49L, Ly-49L, Ly49
Chromosome Location
Chr. 12: 10588478 - 10599835 on Build GRCh38
UniGene ID
Hs.159297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028045.1 5, 6 1040
GenBank mRNA AF047445.1 - 5, 6 770
AK290077.1 - 4, 5 741
AK296103.1 - 5, 6 715
AK304677.1 - 4, 5 736
AK310151.1 - 3, 4 547
BC069672.1 - 5, 6 743 7262588
BC069697.1 - 5, 6 742 7262600
BC069734.1 - 5, 6 743 7262612
BC101471.1 - 757 8068977
BC101473.1 - 760 8068979

View Full Product Details