Gene Transcripts

Gene Symbol
MFAP3
Entrez Gene ID
4238
Gene Name
microfibrillar associated protein 3
Gene Aliases
Chromosome Location
Chr. 5: 154038959 - 154057454 on Build GRCh38
UniGene ID
Hs.432818
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242336.1 NP_001229265.1 1 269
GenBank mRNA AL037715.1 - 1 238
BC026244.1 - 1 229 4697443


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA376590.1 - 1 230 Chr. 5 - 154038999 - 154049772 [+]
GenBankmRNA BG531421.1 - 1 230 Chr. 5 - 154038999 - 154050019 [+]

View Full Product Details