Gene Transcripts

Gene Symbol
TPTEP1
Entrez Gene ID
387590
Gene Name
transmembrane phosphatase with tensin homology pseudogene 1
Gene Aliases
psiTPTE22
Chromosome Location
Chr. 22: 16601911 - 16648830 on Build GRCh38
UniGene ID
Hs.474116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001591.1 4 679
GenBank mRNA AK226145.1 - 2 208
AY219889.1 - 4 679
BC107743.1 - 3 253

View Full Product Details