Gene Transcripts
- Gene Symbol
- LOC202025
- Entrez Gene ID
- 202025
- Gene Name
- uncharacterized LOC202025
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.618463
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL713660.1 |
- |
1 |
670 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000561486.1 |
- |
1 |
666 |
|
Chr. 4 - 10068089 - 10073019 [-] |
View Full Product Details