Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321635.1 | NP_001308564.1 | 11 | 2260 | |
NM_001321636.1 | NP_001308565.1 | 10 | 2165 | ||
NM_024759.2 | NP_079035.1 | 12 | 2288 | ||
NR_135745.1 | 12 | 2424 | |||
NR_135746.1 | 13 | 2546 | |||
NR_135747.1 | 12 | 2484 | |||
GenBank mRNA | AK025015.1 | - | 1 | 57 |