Gene Transcripts
- Gene Symbol
- RRP1
- Entrez Gene ID
- 8568
- Gene Name
- ribosomal RNA processing 1
- Gene Aliases
- D21S2056E, NNP-1, NOP52, RRP1A
- Chromosome Location
- Chr. 21: 43789537 - 43804102 on Build GRCh38
- UniGene ID
- Hs.110757
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW901603.1 |
- |
1 |
452 |
|
Chr. 21 - 43803045 - 43803536 [+] |
GenBankmRNA |
BP263582.1 |
- |
1 |
267 |
|
Chr. 21 - 43803216 - 43803797 [+] |
GenBankmRNA |
BF363881.1 |
- |
1 |
405 |
|
Chr. 21 - 43803083 - 43803555 [+] |
View Full Product Details