Gene Transcripts
- Gene Symbol
- MYO15A
- Entrez Gene ID
- 51168
- Gene Name
- myosin XVA
- Gene Aliases
- DFNB3, MYO15
- Chromosome Location
- Chr. 17: 18108706 - 18180227 on Build GRCh38
- UniGene ID
- Hs.462390
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055873.1 |
- |
1 |
1962 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD512374.1 |
- |
|
201 |
|
Chr. - - [-] |
View Full Product Details