Gene Transcripts
- Gene Symbol
- COL27A1
- Entrez Gene ID
- 85301
- Gene Name
- collagen type XXVII alpha 1
- Gene Aliases
- STLS
- Chromosome Location
- Chr. 9: 114154271 - 114312516 on Build GRCh38
- UniGene ID
- Hs.494892
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI046314.1 |
- |
1 |
163 |
|
Chr. 9 - 114311804 - 114312410 [+] |
GenBankmRNA |
BI494681.1 |
- |
1 |
176 |
|
Chr. 9 - 114311795 - 114312400 [+] |
GenBankmRNA |
CR605514.1 |
- |
1 |
969 |
|
Chr. 9 - 114310997 - 114312467 [+] |
View Full Product Details