Gene Transcripts
- Gene Symbol
- CECR9
- Entrez Gene ID
- 30847
- Gene Name
- cat eye syndrome chromosome region, candidate 9 (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.542791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF307449.1 |
- |
1 |
28 |
|
View Full Product Details