Gene Transcripts
- Gene Symbol
- LOC101928885
- Entrez Gene ID
- 101928885
- Gene Name
- uncharacterized LOC101928885
- Gene Aliases
- Chromosome Location
- Chr. 5: 68967741 - 69029927 on Build GRCh38
- UniGene ID
- Hs.630474
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130778.1 |
|
2 |
384 |
|
GenBank mRNA |
AK128486.1 |
- |
2 |
384 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB212090.1 |
- |
2 |
384 |
|
Chr. 5 - 68970692 - 69029927 [-] |
View Full Product Details