Gene Transcripts
- Gene Symbol
- MEG3
- Entrez Gene ID
- 55384
- Gene Name
- maternally expressed 3 (non-protein coding)
- Gene Aliases
- FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
- Chromosome Location
- Chr. 14: 100826108 - 100861026 on Build GRCh38
- UniGene ID
- Hs.654863
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046473.1 |
|
7 |
4431 |
|
GenBank mRNA |
AK098378.1 |
- |
1 |
1380 |
|
View Full Product Details