Gene Transcripts

Gene Symbol
TMTC1
Entrez Gene ID
83857
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Gene Aliases
ARG99, OLF, TMTC1A
Chromosome Location
Chr. 12: 29500813 - 29784759 on Build GRCh38
UniGene ID
Hs.401954
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020006.1 XP_016875495.1 14 8357
GenBank mRNA AK097137.1 - 1 3375

View Full Product Details