Gene Transcripts
- Gene Symbol
- ERBB4
- Entrez Gene ID
- 2066
- Gene Name
- erb-b2 receptor tyrosine kinase 4
- Gene Aliases
- ALS19, HER4, p180erbB4
- Chromosome Location
- Chr. 2: 211375717 - 212539147 on Build GRCh38
- UniGene ID
- Hs.390729
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024204.1 |
- |
1 |
2119 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DR002628.1 |
- |
1 |
332 |
|
Chr. 2 - 212287134 - 212287579 [-] |
View Full Product Details