Gene Transcripts
- Gene Symbol
- CDRT4
- Entrez Gene ID
- 284040
- Gene Name
- CMT1A duplicated region transcript 4
- Gene Aliases
- Chromosome Location
- Chr. 17: 15436015 - 15467611 on Build GRCh38
- UniGene ID
- Hs.164595
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126240.1 |
- |
1 |
2165 |
|
View Full Product Details