Gene Transcripts

Gene Symbol
LINGO1-AS2
Entrez Gene ID
101929478
Gene Name
LINGO1 antisense RNA 2
Gene Aliases
Chromosome Location
Chr. 15: 77660051 - 77668074 on Build GRCh38
UniGene ID
Hs.569510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120361.1 3 402
NR_120362.1 3 384
GenBank mRNA BC038758.1 - 3 400
BI463387.1 - 3 405
HY007517.1 - 3 373

View Full Product Details