Gene Transcripts
- Gene Symbol
- Gdap6
- Entrez Gene ID
- 14551
- Gene Name
- ganglioside-induced differentiation-associated-protein 6
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.453134
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Y17855.1 |
- |
1 |
183 |
|
View Full Product Details