Gene Transcripts
- Gene Symbol
- Mei4
- Entrez Gene ID
- 75033
- Gene Name
- meiosis-specific, MEI4 homolog (S. cerevisiae)
- Gene Aliases
- 4930486G11Rik
- Chromosome Location
- Chr. 9: 81863647 - 82206007 on Build GRCm38
- UniGene ID
- Mm.335329
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015631.1 |
- |
|
708 |
|
View Full Product Details