Gene Transcripts
- Gene Symbol
- l7Rn6
- Entrez Gene ID
- 67669
- Gene Name
- lethal, Chr 7, Rinchik 6
- Gene Aliases
- 0610007P06Rik, 1110002N09Rik, Hikeshi, l(7)6Rn
- Chromosome Location
- Chr. 7: 89918685 - 89941221 on Build GRCm38
- UniGene ID
- Mm.28679
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_111916.1 |
|
1 |
2013 |
|
GenBank mRNA |
AK133534.1 |
- |
1 |
1972 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI697863.1 |
- |
1 |
38 |
|
Chr. 7 - 89938625 - 89939219 [-] |
View Full Product Details