Gene Transcripts
- Gene Symbol
- Kcnj9
- Entrez Gene ID
- 16524
- Gene Name
- potassium inwardly-rectifying channel, subfamily J, member 9
- Gene Aliases
- 1700085N21Rik, Girk3, Kir3.3, mbGIRK3
- Chromosome Location
- Chr. 1: 172320501 - 172329331 on Build GRCm38
- UniGene ID
- Mm.261168
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA198887.1 |
- |
1 |
23 |
|
Chr. 1 - 172320541 - 172321051 [-] |
GenBankmRNA |
BX517677.1 |
- |
1 |
23 |
|
Chr. 1 - 172320510 - 172321050 [-] |
View Full Product Details