Gene Transcripts
- Gene Symbol
- Gm16208
- Entrez Gene ID
- 102637442
- Gene Name
- predicted gene 16208
- Gene Aliases
- OTTMUSG00000031756
- Chromosome Location
- Chr. 8: 107029411 - 107031192 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028089.1 |
- |
3 |
251 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV207187.2 |
- |
3 |
268 |
|
Chr. 8 - 107029692 - 107031188 [-] |
GenBankmRNA |
BB610313.1 |
- |
3 |
251 |
|
Chr. 8 - 107029660 - 107031188 [-] |
View Full Product Details