Gene Transcripts
- Gene Symbol
- 4930423D24Rik
- Entrez Gene ID
- 73892
- Gene Name
- RIKEN cDNA 4930423D24 gene
- Gene Aliases
- Chromosome Location
- Chr. 10: 117177625 - 117181110 on Build GRCm38
- UniGene ID
- Mm.50199
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015188.1 |
- |
3 |
368 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714739.1 |
- |
4 |
366 |
|
Chr. 10 - 117177629 - 117181114 [+] |
View Full Product Details