Gene Transcripts
- Gene Symbol
- Gm14244
- Entrez Gene ID
- 102641286
- Gene Name
- predicted gene 14244
- Gene Aliases
- OTTMUSG00000016071
- Chromosome Location
- Chr. 2: 127691861 - 127701879 on Build GRCm38
- UniGene ID
- Mm.330572
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029760.1 |
- |
2 |
1068 |
|
View Full Product Details