Gene Transcripts
- Gene Symbol
- Gm19466
- Entrez Gene ID
- 100502945
- Gene Name
- predicted gene, 19466
- Gene Aliases
- Chromosome Location
- Chr. 18: 53648151 - 53705418 on Build GRCm38
- UniGene ID
- Mm.376937
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040349.1 |
|
1 |
195 |
|
GenBank mRNA |
AK044363.1 |
- |
1 |
195 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB642297.1 |
- |
1 |
195 |
|
Chr. 18 - 53648151 - 53703768 [+] |
View Full Product Details