Gene Transcripts
- Gene Symbol
- 5430416G10Rik
- Entrez Gene ID
- 71334
- Gene Name
- RIKEN cDNA 5430416G10 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.461516
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017315.1 |
- |
1 |
1362 |
|
View Full Product Details