Gene Transcripts
- Gene Symbol
- A730082K24Rik
- Entrez Gene ID
- 101786
- Gene Name
- RIKEN cDNA A730082K24 gene
- Gene Aliases
- AU044236
- Chromosome Location
- Chr. 7: 114948650 - 114983967 on Build GRCm38
- UniGene ID
- Mm.395157
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040319.1 |
|
4 |
385 |
|
GenBank mRNA |
AK136021.1 |
- |
4 |
371 |
|
BG082928.2 |
- |
4 |
352 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV648724.1 |
- |
4 |
370 |
|
Chr. 7 - 114951289 - 114975315 [+] |
GenBankmRNA |
CJ052895.1 |
- |
4 |
371 |
|
Chr. 7 - 114951274 - 114975329 [+] |
View Full Product Details