Gene Transcripts
- Gene Symbol
- 1700074G04Rik
- Entrez Gene ID
- 78628
- Gene Name
- RIKEN cDNA 1700074G04 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018899.1 |
- |
1 |
40 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706975.1 |
- |
1 |
40 |
|
Chr. 11 - 116589621 - 116589781 [-] |
View Full Product Details