Gene Transcripts
- Gene Symbol
- 9230110F15Rik
- Entrez Gene ID
- 77080
- Gene Name
- RIKEN cDNA 9230110F15 gene
- Gene Aliases
- Pate-A, mANLP3
- Chromosome Location
- Chr. 9: 35838329 - 35844150 on Build GRCm38
- UniGene ID
- Mm.159846
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020329.1 |
- |
3 |
589 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY721139.1 |
- |
3 |
589 |
|
Chr. 9 - 35838799 - 35844150 [-] |
View Full Product Details