Gene Transcripts
- Gene Symbol
- 4930414N06Rik
- Entrez Gene ID
- 73894
- Gene Name
- RIKEN cDNA 4930414N06 gene
- Gene Aliases
- Chromosome Location
- Chr. 19: 44972117 - 44984550 on Build GRCm38
- UniGene ID
- Mm.491408
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040294.1 |
|
2 |
196 |
|
GenBank mRNA |
AK015139.1 |
- |
2 |
196 |
|
AK076690.1 |
- |
2 |
196 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV260690.2 |
- |
2 |
207 |
|
Chr. 19 - 44972117 - 44984550 [-] |
GenBankmRNA |
BB615011.1 |
- |
2 |
196 |
|
Chr. 19 - 44972113 - 44984550 [-] |
GenBankmRNA |
CN840522.1 |
- |
2 |
176 |
|
Chr. 19 - 44972113 - 44984529 [-] |
GenBankmRNA |
BU935609.1 |
- |
2 |
178 |
|
Chr. 19 - 44960650 - 44984529 [-] |
GenBankmRNA |
BY714687.1 |
- |
|
196 |
|
Chr. - - [-] |
GenBankmRNA |
CA465854.1 |
- |
2 |
191 |
|
Chr. 19 - 44972117 - 44984545 [-] |
View Full Product Details