Gene Transcripts

Gene Symbol
4930517M08Rik
Entrez Gene ID
75117
Gene Name
RIKEN cDNA 4930517M08 gene
Gene Aliases
Chromosome Location
Chr. 17: 4634980 - 4637478 on Build GRCm38
UniGene ID
Mm.429393
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK015816.1 - 3 352


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY715360.1 - 3 352 Chr. 17 - 4634978 - 4637455 [+]

View Full Product Details