Gene Transcripts

Gene Symbol
LINC00632
Entrez Gene ID
286411
Gene Name
long intergenic non-protein coding RNA 632
Gene Aliases
Chromosome Location
Chr. X: 140709759 - 140772677 on Build GRCh38
UniGene ID
Hs.317627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028345.2 2 471
GenBank mRNA BX508035.1 - 2 464
DA128399.1 - 2 471
DA529197.1 - 1 94


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000607004.5 - 2 463 Chr. X - 140709767 - 140712229 [+]

View Full Product Details