Gene Transcripts
- Gene Symbol
- LOC105374266
- Entrez Gene ID
- 105374266
- Gene Name
- uncharacterized LOC105374266
- Gene Aliases
- Chromosome Location
- Chr. 3: 187958775 - 187976407 on Build GRCh38
- UniGene ID
- Hs.570518
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135537.1 |
|
2 |
634 |
|
GenBank mRNA |
BX403058.2 |
- |
2 |
634 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000446091.1 |
- |
2 |
634 |
|
Chr. 3 - 187965768 - 187976407 [-] |
View Full Product Details