Gene Transcripts
- Gene Symbol
- LINC01320
- Entrez Gene ID
- 104355288
- Gene Name
- long intergenic non-protein coding RNA 1320
- Gene Aliases
- Chromosome Location
- Chr. 2: 34677557 - 34722563 on Build GRCh38
- UniGene ID
- Hs.624529
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126404.1 |
|
3 |
948 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000423663.5 |
- |
3 |
926 |
|
Chr. 2 - 34677579 - 34722563 [+] |
View Full Product Details