Gene Transcripts
- Gene Symbol
- GACAT1
- Entrez Gene ID
- 104326057
- Gene Name
- gastric cancer associated transcript 1 (non-protein coding)
- Gene Aliases
- LINC00876
- Chromosome Location
- Chr. 2: 107754112 - 107822129 on Build GRCh38
- UniGene ID
- Hs.585967
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126370.1 |
|
4 |
408 |
|
GenBank mRNA |
DB096621.1 |
- |
4 |
408 |
|
View Full Product Details