Gene Transcripts

Gene Symbol
UBE2E2-AS1
Entrez Gene ID
100505877
Gene Name
UBE2E2 antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 3: 23195070 - 23202993 on Build GRCh38
UniGene ID
Hs.125904
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136176.1 2 402
NR_136177.1 2 426
GenBank mRNA HY029877.1 - 2 405


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000430018.1 - 2 426 Chr. 3 - 23195070 - 23202578 [-]
GENCODE ENST00000452251.5 - 2 414 Chr. 3 - 23195070 - 23202533 [-]
GENCODE ENST00000421375.5 - 2 424 Chr. 3 - 23195070 - 23202543 [-]

View Full Product Details