Gene Transcripts

Gene Symbol
LOC105376351
Entrez Gene ID
105376351
Gene Name
uncharacterized LOC105376351
Gene Aliases
Chromosome Location
Chr. 10: 2446253 - 2501462 on Build GRCh38
UniGene ID
Hs.343482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136146.1 3 808


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000441907.1 - 2 721 Chr. 10 - 2446253 - 2447352 [-]

View Full Product Details