Gene Transcripts
- Gene Symbol
- LINC00837
- Entrez Gene ID
- 100507605
- Gene Name
- long intergenic non-protein coding RNA 837
- Gene Aliases
- Chromosome Location
- Chr. 10: 28789311 - 28796050 on Build GRCh38
- UniGene ID
- Hs.119207
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB524548.1 |
- |
1 |
140 |
|
Chr. 10 - 28789313 - 28789791 [-] |
GENCODE |
ENST00000443246.2 |
- |
2 |
3527 |
|
Chr. 10 - 28789188 - 28796006 [-] |
View Full Product Details