Gene Transcripts
- Gene Symbol
- LOC102723838
- Entrez Gene ID
- 102723838
- Gene Name
- uncharacterized LOC102723838
- Gene Aliases
- Chromosome Location
- Chr. 11: 102467255 - 102498801 on Build GRCh38
- UniGene ID
- Hs.738066
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135076.1 |
|
3 |
462 |
|
GenBank mRNA |
BX093783.1 |
- |
3 |
462 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000529278.1 |
- |
3 |
462 |
|
Chr. 11 - 102467255 - 102498801 [+] |
View Full Product Details