Gene Transcripts
- Gene Symbol
- LOC105369266
- Entrez Gene ID
- 105369266
- Gene Name
- uncharacterized LOC105369266
- Gene Aliases
- Chromosome Location
- Chr. 16: 33329444 - 33331176 on Build GRCh38
- UniGene ID
- Hs.125998
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042588.1 |
- |
1 |
1467 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000566232.1 |
- |
1 |
1633 |
|
Chr. 16 - 32262827 - 32265514 [-] |
View Full Product Details