Gene Transcripts
- Gene Symbol
- LOC152274
- Entrez Gene ID
- 152274
- Gene Name
- uncharacterized LOC152274
- Gene Aliases
- Chromosome Location
- Chr. 3: 14799331 - 14811414 on Build GRCh38
- UniGene ID
- Hs.434128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135245.1 |
|
3 |
1863 |
|
GenBank mRNA |
AK056398.1 |
- |
3 |
1863 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000566553.1 |
- |
3 |
1863 |
|
Chr. 3 - 14799331 - 14811414 [-] |
View Full Product Details